missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Antibody (2B12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-78553-0.025mg
This item is not returnable.
View return policy
Description
Human Serum Albumin Monoclonal specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.
Specifications
| Human Serum Albumin | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| 2B12 | |
| Unconjugated | |
| P02768 | |
| ALB | |
| Human Serum Albumin | |
| 0.025 mg | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| 213 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction