Learn More
Human SHBG Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB26562-SP
 View more versions of this product
Product Code. 18762758
  • 188.00 € / 25µg
Description
SHBG Monoclonal specifically detects SHBG in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SHBG
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ABP, ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin
Mouse myeloma cell line NS0-derived recombinant human SHBG Leu30-His402 Accession # P04278
25 μg
Primary
Detects human SHBG in direct ELISAs.
Human
Purified
ELISA
336729
ELISA
P04278
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
6462
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b