missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SHBG Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB26562-100
This item is not returnable.
View return policy
Description
SHBG Monoclonal specifically detects SHBG in Human samples. It is validated for ELISA.
Specifications
| SHBG | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ABP, ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin | |
| Mouse myeloma cell line NS0-derived recombinant human SHBG Leu30-His402 Accession # P04278 | |
| 100 μg | |
| Primary | |
| Detects human SHBG in direct ELISAs. | |
| Human | |
| Purified |
| ELISA | |
| 336729 | |
| ELISA | |
| P04278 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6462 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction