missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-14 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB4905-100
This item is not returnable.
View return policy
Description
Siglec-14 Monoclonal specifically detects Siglec-14 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Siglec-14 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AC018755.2, sialic acid binding Ig-like lectin 14, Siglec14 | |
| Mouse myeloma cell line, NS0-derived human Siglec-14, Lys18-Leu358, Accession # Q08ET2 | |
| 100 μg | |
| 100049587 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, CyTOF | |
| 2457E | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Q08ET2 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Siglec-14 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction