missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-2/CD22 Antibody, R&D Systems™

Mouse Monoclonal Antibody
196.00 € - 456.00 €
Specifications
| Antigen | Siglec-2/CD22 |
|---|---|
| Clone | 219903 |
| Dilution | Neutralization 6-60 ng/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18763227
![]() |
R&D Systems
MAB19682-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730167
![]() |
R&D Systems
MAB19682-100 |
100 μg |
456.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human samples. It is validated for Neutralization.Tekniske data
| Siglec-2/CD22 | |
| Neutralization 6-60 ng/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P20273 | |
| 933 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 219903 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-2/CD22, Asp20-Arg687, Accession # P20273 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Siglec-2/CD22 in direct ELISAs. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel