missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-3/CD33 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
248.00 € - 458.00 €
Specifications
| Antigen | Siglec-3/CD33 |
|---|---|
| Clone | 6C5/2 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758977
![]() |
R&D Systems
FAB1137A-025 |
25 Tests |
248.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18796746
![]() |
R&D Systems
FAB1137A-100 |
100 Tests |
458.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.Specifications
| Siglec-3/CD33 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 945 | |
| Recombinant human Siglec-3/CD33 Extracellular domain | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 6C5/2 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
| CD33 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Siglec-3/CD33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -5, -7, or -9 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title