missing translation for 'onlineSavingsMsg'
Learn More

Human Siglec-6/CD327 Antibody, R&D Systems™

Product Code. 18012652
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012652 100 μg 100µg
18717027 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18012652 Supplier R&D Systems Supplier No. AF2859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Siglec-6/CD327 Polyclonal specifically detects Siglec-6/CD327 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-6/CD327
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_942142
Gene Alias CD327, CD327 antigen, CD33 antigen-like 1, CD33L1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec6, Siglec-6
Gene Symbols SIGLEC6
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327 Gln27-Val331 Accession # NP_942142
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 946
Test Specificity Detects human Siglec-6/CD327 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Siglec-3, rhSiglec-5 and rhSiglec-9 is observed, approximately 5% cross-reactivity with rhSiglec-10 is observed, and less than 1% cross-reactivity with rhSiglec-2, rhSiglec-7, and rhSiglec-11 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.