missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sirtuin 2/SIRT2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
201.00 € - 592.00 €
Specifications
| Antigen | Sirtuin 2/SIRT2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785757
![]() |
R&D Systems
AF4358-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18090002
![]() |
R&D Systems
AF4358 |
100 μg |
592.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sirtuin 2/SIRT2 Polyclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot.Specifications
| Sirtuin 2/SIRT2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, SIR2L2, SIRT2, sirtuin 2, sirtuin type 2 | |
| SIRT2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8IXJ6 | |
| 22933 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sirtuin 2/SIRT2 Ala2-Gln389 (predicted) Accession # Q8IXJ6 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Sirtuin 2/SIRT2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SIRT1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title