missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC1A5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
261.00 € - 571.00 €
Specifications
| Antigen | SLC1A5 |
|---|---|
| Clone | 1085750 |
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 0.1-10 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245512
|
R&D Systems
MAB11603-SP |
25 μg |
261.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245502
|
R&D Systems
MAB11603-100 |
100 μg |
571.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC1A5 Monoclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Immunohistochemistry,Western Blot,ImmunocytochemistrySpecifications
| SLC1A5 | |
| Western Blot 2 μg/mL, Immunohistochemistry 0.1-10 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| AAAT, Alanine/serine/cysteine transporter 2, ASC amino acid transporter-2, ASCT2, ATB(0), ATBO, baboon M7 virus receptor, M7V1, M7VS1, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRC, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
| Synthetic Peptide, Accession # Q15758 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1085750 | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q15758 | |
| 6510 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title