missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC6A4/5-HTTLPR Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 425.00 €
Specifications
| Antigen | SLC6A4/5-HTTLPR/Serotonin transporter |
|---|---|
| Clone | 632705 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710147
![]() |
R&D Systems
MAB17292-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740817
![]() |
R&D Systems
MAB17292-100 |
100 μg |
425.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC6A4/5-HTTLPR/Serotonin transporter Monoclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human samples. It is validated for Flow CytometrySpecifications
| SLC6A4/5-HTTLPR/Serotonin transporter | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Mouse | |
| Human | |
| P31645 | |
| 6532 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 632705 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5HT transporter, 5-HT Transporter, 5-HTT, 5HTTLPR, 5-HTTLPR, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, Serotonin Transporter 1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
| Human SLC6A4/5-HTTLPR synthetic peptide, Accession # P31645 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title