missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLITRK6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7317-SP
This item is not returnable.
View return policy
Description
SLITRK6 Monoclonal specifically detects SLITRK6 in Human samples. It is validated for Immunocytochemistry.
Specifications
| SLITRK6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK-6 Ile16-Ala604 Accession # Q9H5Y7 | |
| 25 μg | |
| Primary | |
| Detects human SLITRK-6 in direct ELISAs. | |
| Human | |
| IgG2a |
| Immunocytochemistry | |
| 753826 | |
| Immunocytochemistry 10 ug/mL | |
| Q9H5Y7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84189 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction