missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Smoothened Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10276-100
This item is not returnable.
View return policy
Description
Smoothened Monoclonal specifically detects Smoothened in Human samples. It is validated for Immunohistochemistry.
Specifications
| Smoothened | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| bnb, CRJS, FZD11, Gx, Protein Gx, SMO, SMOH, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila) | |
| Chinese Hamster Ovary cell line, CHO-derived human Smoothened, Met1-Tyr233, Accession # Q99835 | |
| 100 μg | |
| Primary | |
| Detects human Smoothened in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 1015418 | |
| Immunohistochemistry 1-25 ug/mL | |
| Q99835 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6608 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction