Learn More
Human Smoothened Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB10276-SP
 View more versions of this product
Product Code. 18760877
  • 209.00 € / 25µg
Description
Smoothened Monoclonal specifically detects Smoothened in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Smoothened
Monoclonal
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
bnb, CRJS, FZD11, Gx, Protein Gx, SMO, SMOH, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila)
Chinese Hamster Ovary cell line, CHO-derived human Smoothened, Met1-Tyr233, Accession # Q99835
25 μg
Primary
Detects human Smoothened in direct ELISAs.
Human
Purified
Immunohistochemistry
1015418
Immunohistochemistry 1-25 ug/mL
Q99835
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
6608
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2a