missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SODD/BAG4 Antibody, R&D Systems™

Description
SODD/BAG4 Polyclonal specifically detects SODD/BAG4 in Human samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.
Specifications
Specifications
| Antigen | SODD/BAG4 |
| Applications | Western Blot, Immunocytochemistry, Immunoassay |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 μg/mL, Immunocytochemistry 5-15 μg/mL, Knockout Validated |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | O95429 |
| Gene Alias | BAG4, BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4 |
| Gene Symbols | BAG4 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?