missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
262.00 € - 547.00 €
Specifications
| Antigen | SOX6 |
|---|---|
| Clone | 667162 |
| Dilution | 5 to 25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796225
![]() |
R&D Systems
MAB77591-100 |
100 μg |
547.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766075
![]() |
R&D Systems
MAB77591-SP |
25 μg |
262.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX6 Monoclonal antibody specifically detects SOX6 in Human samples. It is validated for ImmunohistochemistrySpecifications
| SOX6 | |
| 5 to 25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
| E. coli-derived recombinant human SOX6, Met1-Leu339, Accession # P35712 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 667162 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P35712 | |
| 55553 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title