Learn More
Human SP-D Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB19201-100
 View more versions of this product
Product Code. 18741807
  • 460.00 € / 100µg
Description
SP-D Monoclonal specifically detects SP-D in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SP-D
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Mouse myeloma cell line NS0-derived recombinant human SP-D, Ala21-Phe375, Accession # P35247.2
100 μg
6441
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2b
ELISA
292215
ELISA
P35247.2
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human SP-D in direct ELISAs.
Human
Supernatant