missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ST2/IL-33R Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB5233-100
This item is not returnable.
View return policy
Description
ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot.
Specifications
| ST2/IL-33R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST-2/IL-33 R Lys19-Phe328 Accession # Q01638 | |
| 100 μg | |
| Primary | |
| Detects human ST-2/IL-33R in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 2154D | |
| Western Blot 2 ug/mL | |
| Q01638 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9173 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction