missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STAT5b Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1584
This item is not returnable.
View return policy
Description
STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated.
Specifications
| STAT5b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 2 ug/1 mg cell lysate, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, Knockout Validated | |
| NP_036580 | |
| STAT5B | |
| Human STAT5b synthetic peptide MDSQWIPHAQS Accession # NP_036580 | |
| 100 μg | |
| Primary | |
| Detects human STAT5b in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF, Immunoassay | |
| 389215 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6777 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction