missing translation for 'onlineSavingsMsg'
Learn More

Human Syndecan-1/CD138 Biotinylated Antibody, R&D Systems™

Product Code. 18003062
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18003062 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18003062 Supplier R&D Systems Supplier No. BAF2780
  • ONLINE EXCLUSIVE PRICE
    624.09 €
    552.30 € / 50µg
    Save 71.79 €
    11% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Syndecan-1/CD138 Polyclonal specifically detects Syndecan-1/CD138 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair)∣Immunohistochemistry∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan-1/CD138
Applications ELISA Detection (Matched Antibody Pair), Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. NP_002988
Gene Alias CD138, CD138 antigen, SDC, SDC1, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1, Syndecan1
Gene Symbols SDC1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-1/CD138 Gln18-Glu251 Accession # NP_002988
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6382
Test Specificity Detects human Syndecan-1/CD138 in ELISAs and Western blots. In sandwich ELISAs, less than 0.3% cross-reactivity with recombinant human (rh) Syndecan-2, rhSyndecan-3, rhSyndecan-4,and recombinant mouse Syndecan-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.