missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Testican 2/SPOCK2 Antibody, R&D Systems™

Description
Testican 2/SPOCK2 Monoclonal specifically detects Testican 2/SPOCK2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
| Antigen | Testican 2/SPOCK2 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 327403 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q92563 |
| Gene Alias | FLJ97039, KIAA0275, KIAA0275CWCV, and Kazal-like domains proteoglycan 2, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2, SPOCK2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?