missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TfR2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3120-SP
This item is not returnable.
View return policy
Description
TfR2 Polyclonal specifically detects TfR2 in Human samples. It is validated for Western Blot.
Specifications
| TfR2 | |
| Polyclonal | |
| Western Blot 2 ug/mL, Simple Western 50 ug/mL | |
| Q9UP52 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 7036 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| HFE3, HFE3transferrin receptor protein 2, MGC126368, TfR2, TFRC2, transferrin receptor 2 | |
| Mouse myeloma cell line NS0-derived recombinant human TfR2 Arg105-Phe801 Accession # Q9UP52 | |
| 25 μg | |
| Primary | |
| Detects human TfR2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction