missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Thyroid Whole Tissue Lysate (Adult Whole Tumor)
Shop All Bio Techne ProductsBeskrivelse
The Human Thyroid Whole Tissue Lysate (Adult Whole Tumor) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Tekniske data
Tekniske data
| For Use With (Application) | Western Blot |
| Tissue | Endocrine |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - Human Tumor Tissue: Thyroid |
| Cellular Fraction | Whole |
| Tissue Condition | Tumor |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?