missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIMP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 396.00 €
Specifications
| Antigen | TIMP-3 |
|---|---|
| Clone | 277128 |
| Concentration | LYOPH |
| Dilution | Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Immunoprecipitation, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792208
![]() |
R&D Systems
MAB9731-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18131111
![]() |
R&D Systems
MAB9731 |
100 μg |
396.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-3 Monoclonal specifically detects TIMP-3 in Human samples. It is validated for Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-reported.Specifications
| TIMP-3 | |
| LYOPH | |
| Immunoprecipitation, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P35625 | |
| 7078 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-3 Cys24-Pro211 Accession # P35625 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TIMP-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TIMP-1, -2, or -4 is observed. |
| 277128 | |
| Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| TIMP3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title