missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8999
This item is not returnable.
View return policy
Description
TLR8 Monoclonal specifically detects TLR8 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| TLR8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51311 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 935166 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9NR97 | |
| TLR8 | |
| Chinese hamster ovary cell line CHO-derived recombinant human TLR8 Glu27-Thr827 Accession # Q9NR97 | |
| 100 μg | |
| Primary | |
| Detects human TLR8 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction