missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMED1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2243-SP
This item is not returnable.
View return policy
Description
TMED1 Monoclonal specifically detects TMED1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| TMED1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IL1RL1-Binding Protein, Il1rl1l, IL1RL1LG, IL-1RL1LG, IL1RL1LGIL1RL1-binding protein, Interleukin 1 Receptor-Like 1 Ligand, Interleukin-1 receptor-like 1 ligand, Ly84l, MGC1270, P24g1, P24gamma1, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, Tp24, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1 | |
| Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445 | |
| 25 μg | |
| 11018 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 1009527 | |
| Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 8-25 μg/mL, CyTOF-ready | |
| Q13445 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human TMED1 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction