missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VCAM-1/CD106 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
232.00 € - 527.00 €
Specifications
| Antigen | VCAM-1/CD106 |
|---|---|
| Clone | 1027406 |
| Dilution | Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 15 μg/mL |
| Applications | Immunohistochemistry, Multiplex Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306744
|
R&D Systems
MAB11671-SP |
25 μg |
232.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306749
|
R&D Systems
MAB11671-100 |
100 μg |
527.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
| VCAM-1/CD106 | |
| Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 15 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Chinese Hamster Ovary cell line, CHO-derived human VCAM-1/CD106, Phe25-Glu698, Accession # P19320 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Antibody |
| 1027406 | |
| Immunohistochemistry, Multiplex Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| P19320 | |
| 7412 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title