missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
214.00 € - 477.00 €
Specifications
| Antigen | VEGF |
|---|---|
| Clone | VG1 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready |
| Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730707
![]() |
R&D Systems
MAB2932-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772287
![]() |
R&D Systems
MAB2932-100 |
100 μg |
477.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| VEGF | |
| Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 7422 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| VG1 | |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Recombinant VEGF 189 protein | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title