missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VISTA/B7-H5/PD-1H Antibody, R&D Systems™

Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | VISTA/B7-H5/PD-1H |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 730835 |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q9H7M9 |
| Gene Alias | 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?