missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Vitronectin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2349-SP
This item is not returnable.
View return policy
Description
Vitronectin Monoclonal specifically detects Vitronectin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Vitronectin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT, VTN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7448 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 342603 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| VTN | |
| Human plasma-derived Vitronectin | |
| 25 μg | |
| Primary | |
| Detects human Vitronectin in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant bovine vitronectin or recombinant mouse vitronectin is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction