missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VSIG8 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9418S-100UG
This item is not returnable.
View return policy
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry.
Specifications
| VSIG8 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| V-set and immunoglobulin domain containing 8 | |
| Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13 | |
| 100 μg | |
| 391123 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 961823 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q5VU13 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction