missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ZNF10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9656-100
This item is not returnable.
View return policy
Description
ZNF10 Monoclonal specifically detects ZNF10 in Human samples. It is validated for Western Blot.
Specifications
| ZNF10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| KOX1, KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 | |
| Synthetic peptide containing human ZNF10 Accession # P21506 | |
| 100 μg | |
| 7556 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 980809 | |
| Western Blot 0.5 ug/mL | |
| P21506 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human ZNF10 in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction