missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HuMEC Ready Medium (1X)
GREENER_CHOICE

Product Code. 10776903
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10776903 500 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10776903 Supplier Gibco™ Supplier No. 12752010
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

HuMEC Ready Medium (1X)

Engineered specifically for culture of human mammary epithelial cells, HuMEC Medium is a serum-free formulation with frozen supplements containing growth factor and other components. This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.

Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.

Contents and Storage:
HuMEC Ready Medium consists of HuMEC Basal Serum-Free Medium (12753018) supplemented with HuMEC Supplement Kit (12755013). The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store HuMEC Ready Medium at -5°C to -20°C. Store the supplement mix at -5°C to -20°C.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Quantity 500 mL
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.