missing translation for 'onlineSavingsMsg'
Learn More

(±)-Huperzine A, MedChemExpress

Product Code. 16497318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497318 10 mg 10mg
16439827 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497318 Supplier Medchem Express Supplier No. HY17388
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (±)-Huperzine A, an active Lycopodium alkaloid extracted from traditional Chinese herb, is a potent, selective and reversible acetylcholinesterase (AChE) inhibitor and has been widely used in China for the treatment of Alzheimer's disease (AD).

(±)-Huperzine A, an active Lycopodium alkaloid extracted from traditional Chinese herb, is a potent, selective and reversible acetylcholinesterase (AChE) inhibitor and has been widely used in China for the treatment of Alzheimer's disease (AD).

Specifications

Chemical Name or Material (±)-Huperzine A
Physical Form Solid
CAS 120786-18-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H18N2O
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : ≥ 50 mg/mL (206.34 mM)
SMILES O=C1C=CC(C(/C2=C/C)(N)CC(C)=CC2C3)=C3N1
Molecular Weight (g/mol) 242.32
Formula Weight 242.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.