missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ DMEM/HIGH Powdered Media
Descripción
- High-quality classical media formulations in convenient packaging
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
- Exacting manufacturing processes ensure lot–to–lot consistency
Especificaciones
Especificaciones
| Classification | None |
| Form | Powder |
| Product Type | DMEM/High glucose with L-glutamine, without Sodium Pyruvate |
| Sterilization Method | Non-sterile |
| With Additives | Glutamine, Glucose |
| Without Additives | No Sodium Pyruvate, No Sodium Bicarbonate |
| Quantity | 2 x 5 L |
| Certifications/Compliance | cGMP and ISO 9001:2000 |
| Content And Storage | 2° to 8°C, away from light and moisture |
Seguridad y manipulación
Nombre del producto
- His SpinTrap Kit
- Skin sensitiser Category 1
- Skin corrosion/irritation Category 1 C
- Reproductive toxicity Category 1B
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H360D-May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Nickel; imidazole
- UNKNOWN AQUATIC-Contains 15% of components with unknown hazards to the aquatic environment.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?