missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Composite 5, J.T. Baker™

Product Code. 15544055 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
6 x 1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15544055 6 x 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544055 Supplier JT Baker Supplier No. 8890.1000
  • ONLINE EXCLUSIVE PRICE
    1096.95 €
    251.26 € / 1 L
    Save 845.69 €
    77% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Developed specifically to make water determination easy and trouble-free. J.T. Baker™ HYDRA-POINT™ Composite 5 is a pyridine-free, one-component reagent (Titer: Min. 5mg H2O per mL) for use in Karl Fischer volumetric titrations.

HYDRA-POINT titrants and solvents achieve fast equilibration within the Karl Fischer reaction vessel, giving greater laboratory efficiency and speed of analysis. They are specially formulated for existing application procedures to give consistent, reliable results for all types of samples.

  • Pyridine-free: The use of pyridine in the laboratory has well-recognized hazards, in addition to its unpleasant odor
  • Productivity: Formulated to reach each endpoint quickly, accurately and with reproducibility, enabling more titrations in less time
  • Accuracy & Efficiency: Titers that are more stable with less drift, helping to save time and reduce reagent usage

Specifications

Melting Point 0°C
Boiling Point >34°C
Color Brownish-red
CAS 7446-09-5, 7553-56-2, 288-32-4
Physical Form Liquid
Quantity 6 x 1 L
Molecular Formula C3H4N2
MDL Number MFCD00005183
Packaging Glass bottle
Solubility Information Not or partly miscible
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.08
Concentration or Composition (by Analyte or Components) Imidazole (10-25%), Iodine (5-12.5%), Sulfur dioxide (2.5-10%)
Chemical Name or Material Composite 5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.