missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRA-POINT™ Karl Fischer Coulometric Oven Reagent, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All J.T.Baker ProductsDescripción
- For Coulometric Karl Fischer Titration
Especificaciones
Especificaciones
| Chemical Name or Material | HYDRA-POINT™ Karl Fischer Coulometric Oven Reagent |
| Quantity | 500 mL |
| Flash Point | 21.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Grade | BAKER ANALYZED™ Reagent |
Seguridad y manipulación
Palabra de advertencia
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?