missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Karl Fischer dry methanol (water content max. 0.01%), J.T. Baker™

Product Code. 10114371 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10114371 1 L 1 L
10256131 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10114371 Supplier JT Baker Supplier No. 8898.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric reagents for water determination by Karl Fischer titration

  • Volumetric reagents for water determination by Karl Fischer titration
  • No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
  • They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
  • To be used with HYDRA-POINT™ Composite 2 product no. 8891 or HYDRA-POINT™ Composite 5 product no. 8890

Chemical Identifiers

Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS 67-56-1
Quantity 1 L
Flash Point 11°C
Packaging Glass bottle
UN Number 1230
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.