missing translation for 'onlineSavingsMsg'
Learn More

Hydralazine hydrochloride, MedChemExpress

Product Code. 16448326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448326 100 mg 100mg
16442146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448326 Supplier Medchem Express Supplier No. HYB0464
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hydralazine hydrochloride is a orally active antihypertensive agent, reduces peripheral resistance directly by relaxing the smooth muscle cell layer in arterial vessel. Hydralazine hydrochloride has antioxidant activity, as well as inhibits ROS release and O2 generation with an IC50 value of 9.53 mM and 1.19 mM, respectively.

Hydralazine hydrochloride is a orally active antihypertensive agent, reduces peripheral resistance directly by relaxing the smooth muscle cell layer in arterial vessel. Hydralazine hydrochloride has antioxidant activity, as well as inhibits ROS release and O2 generation with an IC50 value of 9.53 mM and 1.19 mM, respectively.

Specifications

Chemical Name or Material Hydralazine hydrochloride
Physical Form Powder
CAS 304-20-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H9ClN4
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 25 mg/mL (127.14 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (127.14 mM; ultrasonic and adjust pH to 11 with NaOH)
SMILES NNC1=NN=CC2=C1C=CC=C2.Cl
Molecular Weight (g/mol) 196.64
Formula Weight 196.64
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.