missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Benzoic acid, Solstice

Product Code. 17101236
Change view
Click to view available options
Quantity:
6 x 500 g
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17101236 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17101236 Supplier Hydranal Supplier No. 320356X500G
  • ONLINE EXCLUSIVE PRICE
    389.00 €
    362.86 € / Pack of 6
    Save 26.14 €
    6% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buffer substance for Karl Fischer titration

Hydranal™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Benzoic acid
CAS 65-85-0
Quantity 6 x 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.