missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Buffer Acid, Buffer substance for KF titration, Honeywell Fluka™

Product Code. 15684430 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15684430 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684430 Supplier Honeywell-Hydranal Supplier No. 34804500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid buffer medium for Karl Fischer titration in acidic samples (based on imidazole).

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Buffer Acid
Boiling Point 65 °C (1.013 hPa).The physical data is that of the main component.
CAS Min % ≥30.00, ≥5.00
CAS Max % <50.00, <70.00
CAS 7446-09-5, 288-32-4, 67-56-1
Physical Form Liquid
Quantity 500 mL
Flash Point 11°C (51.8°F)
Purity Grade Notes Auxiliaries for KF titration
Packaging Polycoated bottle
UN Number UN3286
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Buffer for Acids
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol, imidazole,sulphur dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.