missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Solstice

Product Code. 15634490
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
500mL
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634490 500 mL 500mL
15624490 1 L 1 L
15654490 6 x 500 mL Pack of 6
15644490 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15634490 Supplier Honeywell-Hydranal Supplier No. 34827500ML
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Composite 1
CAS 288-32-4, 7446-09-5, 111-90-0
Density 1.11g/mL
Quantity 500 mL
Flash Point 90°C (194°F)
compliance-icons
Product Identifier
  • HYDRANAL-Composite 1
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-Ethoxyethoxy)ethanol, imidazole, 2-Methylimidazole, sulphur dioxide, iodine, 1H-Imidazole monoh
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.