missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™

Product Code. 15614450 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
Product Code. Quantity unitSize
15614450 4 x 2.5 L Pack of 4
15624450 500 mL 500mL
15694440 1 L 1 L
15604450 2.5 L 2.5 L
15654450 6 x 500 mL Pack of 6
15644450 6 x 1 L Pack of 6
6 options
This item is not returnable. View return policy

Product Code. 15614450

Brand: Honeywell Fluka 348064X2.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Composite 2
CAS 7446-09-5, 693-98-1, 288-32-4, 111-90-0, 68007-08-9
Density 1.11g/mL
Quantity 4 x 2.5 L
Flash Point 90°C (194°F)
compliance-icons
Product Identifier
  • HYDRANAL-Composite 2
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : imidazole, sulphur dioxide, iodine, 2-Methylimidazole, 1H-Imidazole monohydriodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.