missing translation for 'onlineSavingsMsg'
Learn More

Smart Chemical HYDRANAL™ - Composite 5, Solstice

Product Code. 16204432
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16204432 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16204432 Supplier Hydranal Supplier No. 348051LSC
  • ONLINE EXCLUSIVE PRICE
    222.00 €
    207.25 € / 1 L
    Save 14.75 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Titrating agent for volumetric one-component Karl Fischer titration (methanol free).

Solstice's Smart Chemical Hydranal™ bottles are embedded with an RFID chip, visible in the form of a new label, that captures essential information for every titration, including article code, product name, lot number, shelf life and numeric concentration.

  • Secure Data Transfer: Ensure correct and complete reagent data every time with traceable information flow.
  • Efficient System Setup: Reduce system set-up time with automated reagent data transfer.

The RFID label is currently compatible with Mettler Toledo's Karl Fischer Compact and Excellence line titrators (when upgraded with external RFID reader) and is available on our best-selling Hydranal Volumetric Titrants.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Composite 5
CAS 288-32-4, 68007-08-9, 7553-56-2, 111-90-0, 693-98-1
Density 1.17 g/cm3
Quantity 1 L
Flash Point 99°C
Packaging Mettler-Toledo Smart Chemical Bottle
Grade Karl Fischer
compliance-icons
Product Identifier
  • HYDRANAL Composite 5
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-Ethoxyethoxy)ethanol, sulphur dioxide, 1H-Imidazole monohydriodide, iodine,imidazole, 2-Methyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.