missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 5 K, Solstice

Product Code. 16686163
Change view
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16686163 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16686163 Supplier Hydranal Supplier No. 348166X1LSC
  • ONLINE EXCLUSIVE PRICE
    1175.00 €
    1095.59 € / Pack of 6
    Save 79.41 €
    6% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HYDRANAL™ - Composite 5 K, Titrating agent for volumetric one-comp Karl Fischer titration

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Composite 5 K
CAS Max % <5%
Color Brown
Density 1.170 g/cm3 (20°C)
CAS 111-90-0
Quantity 6 x 1 L
Flash Point 99°C
Synonym DE Solvent; 2-(2-Ethoxyethoxy)ethanol; CARBITOLDE Solvent; 2-(2-Ethoxyethoxy)ethanol; CARBITOL; Diethylene glycol ethyl ether; Ethyldiglycol; Diethylene glycol ethyl ether; Ethyldiglycol
Grade Volumetric One-Component KF Titration
compliance-icons
Product Identifier
  • HYDRANAL NEXTGEN Composite 5K SC
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-Ethoxyethoxy)ethanol, sulphur dioxide, Imidazole, iodine, 2-methylimidazole, 1H-Imidazole monoh
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.