missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AD, Reagent for coulometric KF titration (anolyte solution), for cells without diaphragm, Solstice

Product Code. 15614460
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
500mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15614460 500 mL 500mL
15624460 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15614460 Supplier Honeywell-Hydranal Supplier No. 34810500ML
  • ONLINE EXCLUSIVE PRICE
    248.00 €
    229.05 € / 500mL
    Save 18.95 €
    7% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-56-1

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Coulomat AD
CAS 7446-09-5, 68007-08-9, 288-32-4
Packaging Poly-coated Bottle
Quantity 500 mL
Flash Point 11°C (51.8°F)
UN Number UN1230
Grade Karl Fischer
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Coulomat AD
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol, 2,2'-iminodiethanol; diethanolamine, sulphur dioxide, imidazole, 1H-Imidazole monohydriodi
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.