missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG-Oven, Honeywell Fluka™

Product Code. 17121236 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17121236

Brand: Honeywell Fluka 34739250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Anolyte for coulometric Karl Fischer titration with oven (methanol-based), for cells with and without diaphragm

Hydranal™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat AG-Oven
CAS 57-55-6, 7446-09-5, 288-32-4, 68007-08-9, 67-56-1
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.