missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat CG-K, Reagent for coulometric KF titration in ketones (catholyte solution), Solstice

Product Code. 15604490
Change view
Click to view available options
Quantity:
1 Kit
10 x 5 mL Ampules
Unit Size:
50mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15604490 10 x 5 mL Ampules 50mL
15614490 1 Kit Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15604490 Supplier Honeywell-Hydranal Supplier No. 3482150ML
  • ONLINE EXCLUSIVE PRICE
    313.00 €
    272.38 € / 50mL
    Save 40.62 €
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Catholyte solution formulated for coulometric Karl Fischer titration in ketones.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Coulomat CG-K
CAS 7446-09-5, 288-32-4, 123-39-7
Packaging Glass Bottle
Quantity 10 x 5 mL Ampules
Flash Point 64°C (147.2°F)
Grade Coulometry
compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Coulomat CG-K
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-methylformamide, tetrahydro-2-furylmethanol; tetrahydrofurfuryl alcohol, imidazole, sulphur dioxid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.