missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Kit
10 x 5 mL Ampules
6 x 25 mL
Unit Size:
50mL
Pack of 6
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Coulomat CG |
| CAS | 134227-25-1, 67-56-1, 14426-21-2, 7446-09-5 |
| Quantity | 6 x 25 mL |
| Flash Point | 14°C (57.2°F) |
| UN Number | UN1230 |
Seguridad y manipulación
Nombre del producto
- HYDRANAL-Coulomat CG
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H311 + H331-Toxic in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H370-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Diethanolamine hydrochloride, methanol,Ethanol, 2,2'-iminobis-, hydriodide, sulphur dioxide
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido