missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat E, Reagent for coulometric KF titration (ethanol-based), for cells with and without diaphragm, Honeywell Fluka™

Product Code. p-7122066 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
500mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15664360 500 mL 500mL
15674360 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15664360 Supplier Honeywell-Hydranal Supplier No. 34726500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anolyte for coulometric Karl Fischer titration (ethanol-based), for cells with and without diaphragm

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Coulomat E
CAS 67-56-1, 111-42-2, 64-17-5
Quantity 500 mL
Flash Point 14°C (57.2°F)
Packaging Glass bottle
UN Number UN1993BE
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Coulomat E
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
  • H371-May cause damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,2'-iminodiethanol;diethanolamine, methanol, sulphur dioxide, imidazole, ethanol; ethyl alcohol, 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.