missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - CRM Sodium tartrate dihydrate, Certified reference material for Karl Fischer titration (water content ∼15.66%), Solstice

Product Code. 15675725
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15675725 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15675725 Supplier Honeywell-Hydranal Supplier No. 3442410G
 Restricted Item
Estimated Shipment: 10-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified Reference Material for Karl Fischer titration (water content ∼15.66%, exact value on certificate of analysis)

Advantages of HYDRANAL- CRM Water Standards:

  • For volumetric and coulometric Karl Fischer applications
  • Manufactured according to ISO 17034 requirements
  • Certificate of Analysis showing the exact water content, expanded uncertainty plus comprehensive product information and instruction for use
  • Lot-related stability and homogeneity studies
  • Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
  • Convenient packaging
TRUSTED_SUSTAINABILITY

Specifications

CAS 6106-24-7
Density 1.820 g/cm3
Quantity 10 g
CAS Max % ≤100.0000%
Molecular Formula C4H8Na2O8
MDL Number MFCD00150035
Packaging Glass Bottle
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Molecular Weight (g/mol) 230.08
PubChem CID 131855972
Grade Certified Reference Material
Assay effective water volumetric
Chemical Name or Material HYDRANAL™ - CRM Sodium tartrate dihydrate, Certified reference material for Karl Fischer titration (water content ∼15.66%)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.